EMAIL THIS PAGE TO A FRIEND
Cell stem cell
Efficient generation of human iPSCs by a synthetic self-replicative RNA.
PMID 23910086
Abstract
The generation of human induced pluripotent stem cells (iPSCs) holds great promise for the development of regenerative medicine therapies to treat a wide range of human diseases. However, the generation of iPSCs in the absence of integrative DNA vectors remains problematic. Here, we report a simple, highly reproducible RNA-based iPSC generation approach that utilizes a single, synthetic self-replicating VEE-RF RNA replicon that expresses four reprogramming factors (OCT4, KLF4, and SOX2, with c-MYC or GLIS1) at consistent high levels prior to regulated RNA degradation. A single VEE-RF RNA transfection into newborn or adult human fibroblasts resulted in efficient generation of iPSCs with all thexa0hallmarks of stem cells, including cell surface markers, global gene expression profiles, and inxa0vivo pluripotency, to differentiate into all three germ layers. The VEE-RF RNA-based approach has broad applicability for the generation of iPSCs for ultimate use in human stem cell therapies in regenerative medicine.