EMAIL THIS PAGE TO A FRIEND
Nutrients
Magnesium and osteoporosis: current state of knowledge and future research directions.
PMID 23912329
Abstract
A tight control of magnesium homeostasis seems to be crucial for bone health. On the basis of experimental and epidemiological studies, both low and high magnesium have harmful effects on the bones. Magnesium deficiency contributes to osteoporosis directly by acting on crystal formation and on bone cells and indirectly by impacting on the secretion and the activity of parathyroid hormone and by promoting low grade inflammation. Less is known about the mechanisms responsible for the mineralization defects observed when magnesium is elevated. Overall, controlling and maintaining magnesium homeostasis represents a helpful intervention to maintain bone integrity.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
GF86050199
Magnesium, microfoil, 25x25mm, thinness 1.0μm, specific density 174μg/cm2, 6 micron aluminum permanent support, 99.9%
Mg
GF67975172
Magnesium, wire reel, 2m, diameter 1.1mm, as drawn, 99.9+%
Mg
GF80461964
Magnesium, microleaf, 25x25mm, thinness 1.0μm, specific density 174μg/cm2, removable support, 99.9%
Mg
GF57746166
Magnesium, microleaf, 50x50mm, thinness 1.0μm, specific density 174μg/cm2, removable support, 99.9%
Mg
GF18270637
Magnesium, wire reel, 0.1m, diameter 0.125mm, as drawn, 99.9+%
Mg
GF96495609
Magnesium, wire reel, 0.1m, diameter 0.25mm, as drawn, 99.9+%
Mg
GF04987547
Magnesium, wire reel, 0.1m, diameter 0.4mm, as drawn, 99.9+%
Mg
GF23637371
Magnesium, wire reel, 0.1m, diameter 0.5mm, as drawn, 99.9+%
Mg
GF83427745
Magnesium, wire reel, 0.1m, diameter 1.0mm, as drawn, 99.9+%
Mg
GF86178189
Magnesium, wire reel, 0.1m, diameter 1.1mm, as drawn, 99.9+%
Mg
GF83829178
Magnesium, wire reel, 0.5m, diameter 0.125mm, as drawn, 99.9+%
Mg
GF61950618
Magnesium, wire reel, 0.5m, diameter 0.25mm, as drawn, 99.9+%
Mg
GF23443646
Magnesium, wire reel, 0.5m, diameter 0.4mm, as drawn, 99.9+%
Mg
GF54671924
Magnesium, wire reel, 0.5m, diameter 0.5mm, as drawn, 99.9+%
Mg
GF28467636
Magnesium, wire reel, 0.5m, diameter 1.0mm, as drawn, 99.9+%
Mg
GF72273157
Magnesium, wire reel, 0.5m, diameter 1.1mm, as drawn, 99.9+%
Mg
GF78647293
Magnesium, wire reel, 1m, diameter 0.125mm, as drawn, 99.9+%
Mg
GF43827071
Magnesium, wire reel, 1m, diameter 0.25mm, as drawn, 99.9+%
Mg
GF30456053
Magnesium, wire reel, 1m, diameter 0.4mm, as drawn, 99.9+%
Mg
GF33990598
Magnesium, wire reel, 1m, diameter 0.5mm, as drawn, 99.9+%
Mg
GF58454983
Magnesium, wire reel, 1m, diameter 1.0mm, as drawn, 99.9+%
Mg
GF34143803
Magnesium, wire reel, 1m, diameter 1.1mm, as drawn, 99.9+%
Mg
GF30911778
Magnesium, wire reel, 2m, diameter 0.125mm, as drawn, 99.9+%
Mg
GF78384437
Magnesium, wire reel, 2m, diameter 0.25mm, as drawn, 99.9+%
Mg
GF07006328
Magnesium, wire reel, 2m, diameter 0.4mm, as drawn, 99.9+%
Mg
GF48824208
Magnesium, wire reel, 2m, diameter 0.5mm, as drawn, 99.9+%
Mg
GF06872924
Magnesium, wire reel, 2m, diameter 1.0mm, as drawn, 99.9+%
Mg
GF56273777
Magnesium, microfoil, 50x50mm, thinness 1.0μm, specific density 174μg/cm2, 6 micron aluminum permanent support, 99.9%
Mg