EMAIL THIS PAGE TO A FRIEND
Journal of magnetic resonance (San Diego, Calif. : 1997)
Para-hydrogen perspectives in hyperpolarized NMR.
PMID 23932399
Abstract
The first instance of para-hydrogen induced polarization (PHIP) in an NMR experiment was serendipitously observed in the 1980s while investigating a hydrogenation reaction (Seldler et al., 1983; Bowers and Weitekamp, 1986, 1987; Eisenschmid et al., 1987) [1-4]. Remarkably a theoretical investigation of the applicability of para-hydrogen as a hyperpolarization agent was being performed in the 1980's thereby quickly providing a theoretical basis for the PHIP-effect (Bowers and Weitekamp, 1986) [2]. The discovery of signal amplification by a non-hydrogenating interaction with para-hydrogen has recently extended the interest to exploit the PHIP effect, as it enables investigation of compounds without structural alteration while retaining the advantages of spectroscopy with hyperpolarized compounds [5]. In this article we will place more emphasis of the future applications of the method while only briefly discussing the efforts that have been made in the understanding of the phenomenon and the development of the method so far.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
00473
Hydrogen, ≥99.999%
H2