PloS one
Characterization of leucocin B-KM432Bz from Leuconostoc pseudomesenteroides isolated from boza, and comparison of its efficiency to pediocin PA-1.
PMID 23936441
Abstract
A bacteriocin-producing bacterium was isolated from boza and identified as Leuconostoc pseudomesenteroides KM432Bz. The antimicrobial peptide was purified and shown to be identical to other class IIa bacteriocins: leucocin A from Leuconostoc gelidum UAL-187 and Leuconostoc pseudomesenteroides QU15 and leucocin B from Leuconostoc carnosum Ta11a. The bacteriocin was named leucocin B-KM432Bz. Leucocin B-KM432Bz gene cluster encodes the bacteriocin precursor (lcnB), the immunity protein (lcnI) and the dedicated export machinery (lcnD and lcnE). A gene of unknown and non-essential function (lcnC), which is interrupted by an insertion sequence of the IS30 family, is localized between lcnB and lcnD. The activity of leucocin B-KM432Bz requires subunit C of the EII(t) Man mannose permease, which is the receptor for entry into target cells. The determination of the minimum inhibitory concentrations revealed the lowest values for leucocin B-KM432Bz over Listeria strains, with 4 to 32 fold better efficiency than pediocin PA-1.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
V3204
4-Vinylpyridine, contains 100 ppm hydroquinone as inhibitor, 95%
C7H7N