EMAIL THIS PAGE TO A FRIEND
BMC veterinary research
Pharmacokinetics of flunixin meglumine in mature swine after intravenous, intramuscular and oral administration.
PMID 23941181
Abstract
The purpose of this study was to determine intravenous (IV), intramuscular (IM) and oral (PO) FM PK in mature swine. Appropriate pain management for lameness in swine is a critical control point for veterinarians and producers, but science-based guidance on optimal housing, management and treatment of lameness is deficient. Six mature swine (121-168xa0kg) were administered an IV, IM, or PO dose of flunixin meglumine at a target dose of 2.2xa0mg/kg in a cross-over design with a 10xa0day washout period between treatments. Plasma samples collected up to 48xa0hours post-administration were analyzed by high pressure liquid chromatography and mass spectrometry (HPLC-MS) followed by non-compartmental pharmacokinetic analysis. No adverse effects were observed with flunixin meglumine administration for all routes. Flunixin meglumine was administered at an actual mean dose of 2.21xa0mg/kg (range: 2.05-2.48xa0mg/kg) IV, IM and PO. A mean peak plasma concentration (CMAX) for IM and PO administration was 3748xa0ng/ml (range: 2749-6004xa0ng/ml) and 946xa0ng/ml (range: 554-1593xa0ng/ml), respectively. TMAX was recorded at 1.00xa0hour (range: 0.50-2.00xa0hours) and 0.61xa0hours (range: 0.17-2.00xa0hours) after PO and IM administration. Half-life (T ½ λz) for IV, IM and PO administration was 6.29xa0hours (range: 4.84-8.34xa0hours), 7.49xa0hours (range: 5.55-12.98xa0hours) and 7.08xa0hours (range: 5.29-9.15xa0hours) respectively. In comparison, bioavailability (F) for PO administration was 22% (range: 11-44%) compared to IM F at 76% (range: 54-92%). The results of the present study suggest that FM oral administration is not the most effective administration route for mature swine when compared to IV and IM. Lower F and Cmax of PO-FM in comparison to IM-FM suggest that PO-FM is less likely to be an effective therapeutic administration route.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
33586
Flunixin, VETRANAL, analytical standard
C14H11F3N2O2