EMAIL THIS PAGE TO A FRIEND
Nihon Naika Gakkai zasshi. The Journal of the Japanese Society of Internal Medicine
[The cutting-edge of medicine; hyponatremia and water metabolism disorders in elderly].
PMID 23947246