EMAIL THIS PAGE TO A FRIEND
Journal of the American Chemical Society
Bioconjugation by native chemical tagging of C-H bonds.
PMID 23957305
Abstract
A general C-H functionalization method for the tagging of natural products and pharmaceuticals is described. An azide-containing sulfinate reagent allows the appendage of azidoalkyl chains onto heteroaromatics, the product of which can then be attached to a monoclonal antibody by a "click" reaction. This strategy expands the breadth of bioactive small molecules that can be linked to macromolecules in a manner that is beyond the scope of existing methods in bioconjugation to permit tagging of the "seemingly untaggable".
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
746118
Sodium difluoroheptylazidosulfinate, 95%
C7H12F2N3NaO2S