EMAIL THIS PAGE TO A FRIEND
OncoTargets and therapy
Adjuvant chemotherapy for luminal A breast cancer: a prospective study comparing two popular chemotherapy regimens.
PMID 23966793
Abstract
Based on the variable benefit of taxanes in the adjuvant setting of early breast cancer in certain tumor phenotypes, especially in human epidermal growth factor receptor (HER)2-positive and triple-negative disease, and with the observation of a lesser benefit in luminal A, this research article aimed at exploring the value of docetaxel in patients with an estrogen receptor-positive, HER2-negative disease phenotype, who might not derive the same benefits as those with other phenotypes. This was a randomized prospective study comparing disease-free survival (DFS) and safety profile of sequential adjuvant three cycles Fluorouracil, Epirubicin, Cyclophosphamide followed by three cycles Docetaxel (FEC-D) versus six cycles classic Fluorouracil, Epirubicin, Cyclophosphamide (FEC)-100 in 60 Egyptian women who presented to Dar Al Fouad Hospital during the period June 2007 to July 2008 with (pT1-2 pN0-3 M0). The primary end point was DFS in a follow-up period of 4 years. The secondary end point was toxicity profile. Four-year DFS rates were comparable in both arms: 73.3% ± 8.1% in the FEC-D arm versus 76.5% ± 7.8% in the FEC-100 arm (P = 0.83). N3 and grade III subgroups achieved the worst DFS in both subgroups (P = 0.001 and P = 0.214, respectively). The rate of nausea and vomiting was higher in the FEC-100 arm (P = 0.49), while grade III-IV neutropenia and febrile neutropenia incidence was similar between both arms. Sequential adjuvant chemotherapy with FEC followed by docetaxel achieved comparable DFS results to FEC alone in luminal A phenotype subgroups of breast cancer.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
757349
Fluoroethylene carbonate, 99%
C3H3FO3