EMAIL THIS PAGE TO A FRIEND
Cell calcium
Human TRPV5 and TRPV6: key players in cadmium and zinc toxicity.
PMID 23968883
Abstract
TRPV5 and TRPV6 are two major calcium transport pathways in the human body maintaining calcium homeostasis. TRPV5 is mainly expressed in the distal convoluted and connecting tubule where it is the major, regulated pathway for calcium reabsorption. TRPV6 serves as an important calcium entry pathway in the duodenum and the placenta. Previously, we showed that human TRPV6 (hTRPV6) transports several heavy metals. In this study we tested whether human TRPV5 (hTRPV5) also transports cadmium and zinc, and whether hTRPV5 together with hTRPV6 are involved in cadmium and zinc toxicity. The hTRPV5 mRNA and protein were expressed in HEK293 cells transiently transfected with pTagRFP-C1-hTRPV5. The overexpression of the hTRPV5 protein at the plasma membrane was revealed by cell surface biotinylation and immunofluorescence techniques. We observed that both cadmium and zinc permeate hTRPV5 in ion imaging experiments using Fura-2 or Newport Green DCF. Our results were further confirmed using whole-cell patch clamp technique. Transient overexpression of hTRPV5 or hTRPV6 sensitized cells to cadmium and zinc. Toxicity curves of cadmium and zinc were also shifted in hTRPV6 expressing HEK293 cells clones. Our results suggest that TRPV5 and TRPV6 are crucial gates controlling cadmium and zinc levels in the human body especially under low calcium dietary conditions, when these channels are maximally upregulated.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
AV49458
Anti-TRPV5 antibody produced in rabbit, affinity isolated antibody