EMAIL THIS PAGE TO A FRIEND
Natural product communications
Carvone as a versatile chiral building block for total syntheses of heterocyclic sesquiterpenoids.
PMID 23980427
Abstract
This review article is focused on the total syntheses of heterocyclic sesquiterpenoids starting from carvone since 1996.