EMAIL THIS PAGE TO A FRIEND
Nanoscale
Gold nanoparticles and fluorescently-labelled DNA as a platform for biological sensing.
PMID 23982570
Abstract
In the past decade gold nanoparticle-nucleic acid conjugates became progressively important for biomedical applications. Fluorophores attached to nucleic acid-gold nanoparticle conjugates have opened up a new era of biological sensing. The most promising advancement in this field was the invention of the so-called 'nano-flare' systems. These systems are capable of detecting specific endocellular targets such as mRNAs, microRNAs or small molecules in real time. In this minireview, we discuss the current progress in the field of DNA-nanoparticles as sensors, their properties, stability, cellular uptake and cytotoxicity.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
326542
Gold, beads, 1-6 mm, 99.999% trace metals basis
Au
373168
Gold, evaporation slug, diam. × L 0.6 cm × 1.2 cm, 99.99% trace metals basis
Au
373176
Gold, evaporation slug, diam. × L 0.6 cm × 0.6 cm, 99.99% trace metals basis
Au
373184
Gold, evaporation slug, diam. × L 0.3 cm × 0.6 cm, 99.99% trace metals basis
Au
265829
Gold, foil, thickness 0.5 mm, 99.99% trace metals basis
Au
349240
Gold, foil, thickness 0.25 mm, ≥99.9% trace metals basis
Au
326496
Gold, foil, thickness 0.127 mm, 99.99% trace metals basis
Au
265810
Gold, foil, thickness 0.1 mm, 99.99% trace metals basis
Au
349275
Gold, foil, thickness 0.05 mm, 99.99% trace metals basis
Au
268461
Gold, foil, thickness 0.025 mm, 99.99% trace metals basis
Au
349267
Gold, foil, thickness 0.025 mm, ≥99.9% trace metals basis
Au
255734
Gold, powder, <850 μm, ≥99.99% trace metals basis
Au
265772
Gold, powder, <45 μm, 99.99% trace metals basis
Au
718424
Gold, powder, size 10-20 μm
Au
326585
Gold, powder, <10 μm, ≥99.9% trace metals basis
Au
636347
Gold, nanopowder, <100 nm particle size, 99.9% trace metals basis
Au
265837
Gold, rod, diam. 3.0 mm, 99.99% trace metals basis
Au
265802
Gold, wire, diam. 2.0 mm, 99.99% trace metals basis
Au
349291
Gold, wire, diam. 1.5 mm, 99.999% trace metals basis
Au
349305
Gold, wire, diam. 1.0 mm, 99.997% trace metals basis
Au
265799
Gold, wire, diam. 1.0 mm, 99.99% trace metals basis
Au
349313
Gold, wire, diam. 0.5 mm, 99.999% trace metals basis
Au
310980
Gold, wire, diam. 0.5 mm, 99.99% trace metals basis
Au
265780
Gold, wire, diam. 0.25 mm, 99.99% trace metals basis
Au
326534
Gold, wire, diam. 0.25 mm, ≥99.9% trace metals basis
Au
326526
Gold, wire, diam. 0.127 mm, 99.99% trace metals basis
Au
349283
Gold, wire, diam. 0.1 mm, 99.99% trace metals basis
Au
GF43139538
Gold, insulated wire, 5m, conductor diameter 0.025mm, insulation thickness 0.005mm, polyester insulation, 99.99%
Au
GF92307922
Gold, insulated wire, 5m, conductor diameter 0.05mm, insulation thickness 0.007mm, polyester insulation, 99.99%
Au
GF11444611
Gold, insulated wire, 5m, conductor diameter 0.05mm, insulation thickness 0.007mm, polyimide insulation, 99.99%
Au