EMAIL THIS PAGE TO A FRIEND
Organic letters
Construction of multifunctional modules for drug discovery: synthesis of novel thia/oxa-azaspiro[3.4]octanes.
PMID 23991674
Abstract
New classes of thia/oxa-azaspiro[3.4]octanes are synthesized through the implementation of robust and step-economic routes. The targeted spirocycles have been designed to act as novel, multifunctional, and structurally diverse modules for drug discovery. Furthermore, enantioselective approaches to the spirocycles are reported.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
O6379
Oxytocin acetate salt hydrate, ≥97% (HPLC)
C43H66N12O12S2