EMAIL THIS PAGE TO A FRIEND
Oncogene
Activation of Akt signaling in prostate induces a TGFβ-mediated restraint on cancer progression and metastasis.
PMID 23995785
Abstract
Mutations in the PTEN tumor suppressor gene are found in a high proportion of human prostate cancers, and in mice, Pten deletion induces high-grade prostate intraepithelial neoplasia (HGPIN). However, progression from HGPIN to invasive cancer occurs slowly, suggesting that tumorigenesis is subject to restraint. We show that Pten deletion, or constitutive activation of the downstream kinase AKT, activates the transforming growth factor (TGF)β pathway in prostate epithelial cells. TGFβ signaling is known to have a tumor suppressive role in many cancer types, and reduced expression of TGFβ receptors correlates with advanced human prostate cancer. We demonstrate that in combination either with loss of Pten or expression of constitutively active AKT1, inactivation of TGFβ signaling by deletion of the TGFβ type II receptor gene relieves a restraint on tumorigenesis. This results in rapid progession to lethal prostate cancer, including metastasis to lymph node and lung. In prostate epithelium, inactivation of TGFβ signaling alone is insufficient to initiate tumorigenesis, but greatly accelerates cancer progression. The activation of TGFβ signaling by Pten loss or AKT activation suggests that the same signaling events that have key roles in tumor initiation also induce the activity of a pathway that restrains disease progression.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
AB3849
Anti-Smad2 Antibody, phospho-specific (Ser465/467), 100 µL, Use Anti-Smad2 Antibody, phospho-specific (Ser465/467) (Rabbit Polyclonal Antibody) validated in ICC, IF, IHC(P), WB to detect Smad2 also known as MAD-mothers against decapentaplegic-homolog 2.
SAB1405987
Anti-CFI antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
HPA001143
Anti-CFI antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
HPA024061
Anti-CFI antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB1410687
Anti-CFI antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
SAB1405869
Anti-GIF antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
HPA039908
Anti-GIF antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
HPA040774
Anti-GIF antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB1410525
Anti-GIF antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
WH0003426M1
Monoclonal Anti-IF antibody produced in mouse, clone 1B3, purified immunoglobulin, buffered aqueous solution