EMAIL THIS PAGE TO A FRIEND
Tetrahedron
Chemoselective hydrosilylation of hydroxyketones.
PMID 23997314
Abstract
A chemoselective method for the hydrosilylation of ketones has been developed, using the combination of triphenylsilane and a catalyst prepared from Ni(COD)2 and the simple N-heterocyclic carbene IMes. The most notable feature of this method is that free hydroxyls are largely unaffected, thus providing a simple one-step procedure for the conversion of hydroxyketones to mono-protected diols, wherein the protecting group is exclusively installed on the ketone-derived hydroxyl. The process is typically high yielding with both simple ketones and more complex hydroxyketone substrates.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
423807
5-Hydroxy-2-adamantanone, 98%
C10H14O2