EMAIL THIS PAGE TO A FRIEND
Environmental pollution (Barking, Essex : 1987)
Identification of novel micropollutants in wastewater by a combination of suspect and nontarget screening.
PMID 24012788
Abstract
To detect site-specific, suspected and formerly unknown contaminants in a wastewater treatment plant effluent, we established a screening procedure based on liquid chromatography-high resolution mass spectrometry (LC-HRMS) with stepwise identification schemes. Based on automated substructure searches a list of 2160 suspected site-specific and documented water contaminants was reduced to those amenable to LC-HRMS. After searching chromatograms for exact masses of suspects, presumably false positive detections were stepwise excluded by retention time prediction, the evaluation of isotope patterns, ionization behavior, and HRMS/MS spectra. In nontarget analysis, peaks for identification were selected based on distinctive isotope patterns and intensity. The stepwise identification of nontarget compounds was automated by a plausibility check of molecular formulas using the Seven Golden Rules, an exclusion of compounds with presumably low commercial importance and an automated HRMS/MS evaluation. Six suspected and five nontarget chemicals were identified, of which two have not been previously reported as environmental pollutants.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
454451
2-(2-Benzothiazolylthio)ethanol, 95%
C9H9NOS2