EMAIL THIS PAGE TO A FRIEND
Scientific reports
Super-paramagnetic responsive nanofibrous scaffolds under static magnetic field enhance osteogenesis for bone repair in vivo.
PMID 24030698
Abstract
A novel nanofibrous composite scaffold composed of super-paramagnetic γ-Fe2O3 nanoparticles (MNP), hydroxyapatite nanoparticles (nHA) and poly lactide acid (PLA) was prepared using electrospinning technique. The scaffold well responds extern static magnetic field with typical saturation magnetization value of 0.049 emu/g as well as possesses nanofibrous architecture. The scaffolds were implanted in white rabbit model of lumbar transverse defects. Permanent magnets are fixed in the rabbit cages to provide static magnetic field for the rabbits post surgery. Results show that MNP incorporated in the nanofibers endows the scaffolds super-paramagnetic responsive under the applied static magnetic field, which accelerates new bone tissue formation and remodeling in the rabbit defect. The scaffold also exhibits good compatibility of CK, Cr, ALT and ALP within normal limits in the serum within 110 days post implantation. In conclusion, the super-paramagnetic responding scaffold with applying of external magnetic field provides a novel strategy for scaffold-guided bone repair.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
P5178
Phenobarbital sodium salt
C12H11N2NaO3