EMAIL THIS PAGE TO A FRIEND
Hepatology research : the official journal of the Japan Society of Hepatology
Splenic lymph follicles generate immunoglobulin M-producing B cells in primary biliary cirrhosis.
PMID 24033874
Abstract
To reveal the site of immunoglobulin (Ig)M production in primary biliary cirrhosis (PBC) we performed immunohistochemical analysis on spleens collected from patients with PBC. Splenic tissue samples were collected at the time of the autopsy from patients with hepatic failure. Immunostaining for IgM, CD21 and CXCL13 were performed using the splenic tissue samples. The samples from five out of eight cases with PBC but not in eight cases of chronic hepatitis C virus infection showed accumulation of IgM positive cells in CD21 positive lymph follicles. The CXCL13 positive cells also accumulated in the center of the lymph follicles where the IgM positive cells accumulated. The present results suggest that excess IgM is produced from the spleen of PBC. Furthermore, it was suggested that CXCL13 positive follicular dendritic cells possibly contribute to this process.