EMAIL THIS PAGE TO A FRIEND
International journal of cancer
Decreased TIP30 expression predicts poor prognosis in pancreatic cancer patients.
PMID 24037692
Abstract
Pancreatic ductal adenocarcinoma (PDAC) is known for its aggressive growth, and is characterized by early tissue invasion and metastasis with poor prognosis. Identifying prognostic markers and delineating the underlying mechanisms that promote progression of PDAC are important for the treatment of pancreatic cancer. TIP30, a newly identified tumor suppressor, appears to be involved in multiple processes during tumor development and metastasis. Here, we investigated the expression of TIP30 in PDAC and its prognostic value in PDAC patients. We examined the expression of TIP30 by immunohistochemistry in tissue microarrays containing 106 surgically resected PDAC. Kaplan-Meier analysis and Cox proportional hazards regression modeling analysis showed that TIP30 expression independently predicted better survival in pancreatectomy patients (p < 0.01). Moreover, decreased TIP30 expression was associated with lymph node metastasis (p < 0.05) and loss of E-cadherin expression (r = 0.329, p < 0.01). Suppression of TIP30 resulted in upregulation of Snail and subsequent downregulation of E-cadherin in SW1990 cells containing high-level of endogenous TIP30. However, in the PANC-1 cells containing low level of endogenous TIP30, suppressing TIP30 caused upregulation of Slug instead of Snail, followed by upregulation of MMP9 rather than E-cadherin. Taken together, our work reveals that decreased TIP30 expression is able to enhance invasion and metastasis of pancreatic cancer cells through upregulation of the Snail family members and may serve as an independent predictor for poor outcomes in PDAC patients.