EMAIL THIS PAGE TO A FRIEND
Angewandte Chemie (International ed. in English)
Arylation of diazoesters by a transient N-H insertion organocascade.
PMID 24038873
Abstract
It takes two: A unique organocatalyzed cascade for the unsymmetric double arylation of α-nitrodiazoesters is described. This organocascade features the strategic use of carbene-activating anilines in conjunction with a urea catalyst, thus allowing for the synthesis of pharmaceutically attractive α-diarylesters through a transient NH insertion process.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
803871
Mattson Boronate Urea Pinacol Ester
C21H21BF6N2O3
804452
Potassium 3-ethoxycarbonylbenzoyltrifluoroborate
C10H9BF3KO3