PloS one
Patterning in placental 11-B hydroxysteroid dehydrogenase methylation according to prenatal socioeconomic adversity.
PMID 24040322
Abstract
Prenatal socioeconomic adversity as an intrauterine exposure is associated with a range of perinatal outcomes although the explanatory mechanisms are not well understood. The development of the fetus can be shaped by the intrauterine environment through alterations in the function of the placenta. In the placenta, the HSD11B2 gene encodes the 11-beta hydroxysteroid dehydrogenase enzyme, which is responsible for the inactivation of maternal cortisol thereby protecting the developing fetus from this exposure. This gene is regulated by DNA methylation, and this methylation and the expression it controls has been shown to be susceptible to a variety of stressors from the maternal environment. The association of prenatal socioeconomic adversity and placental HSD11B2 methylation has not been examined. Following a developmental origins of disease framework, prenatal socioeconomic adversity may alter fetal response to the postnatal environment through functional epigenetic alterations in the placenta. Therefore, we hypothesized that prenatal socioeconomic adversity would be associated with less HSD11B2 methylation. We examined the association between DNA methylation of the HSD11B2 promoter region in the placenta of 444 healthy term newborn infants and several markers of prenatal socioeconomic adversity: maternal education, poverty, dwelling crowding, tobacco use and cumulative risk. We also examined whether such associations were sex-specific. We found that infants whose mothers experienced the greatest levels of socioeconomic adversity during pregnancy had the lowest extent of placental HSD11B2 methylation, particularly for males. Associations were maintained for maternal education when adjusting for confounders (p<0.05). Patterns of HSD11B2 methylation suggest that environmental cues transmitted from the mother during gestation may program the developing fetus's response to an adverse postnatal environment, potentially via less exposure to cortisol during development. Less methylation of placental HSD11B2 may therefore be adaptive and promote the effective management of stress associated with social adversity in a postnatal environment.
Related Materials