EMAIL THIS PAGE TO A FRIEND
Journal of neurochemistry
c-Jun N-terminal kinase regulates mGluR-dependent expression of post-synaptic FMRP target proteins.
PMID 24047560
Abstract
Fragile X syndrome (FXS) is caused by the loss of functional fragile X mental retardation protein (FMRP). Loss of FMRP results in an elevated basal protein expression profile of FMRP targeted mRNAs, a loss of local metabotropic glutamate receptor (mGluR)-regulated protein synthesis, exaggerated long-term depression and corresponding learning and behavioral deficits. Evidence shows that blocking mGluR signaling in FXS models ameliorates these deficits. Therefore, understanding the signaling mechanisms downstream of mGluR stimulation may provide additional therapeutic targets for FXS. Kinase cascades are an integral mechanism regulating mGluR-dependent protein translation. The c-Jun N-terminal kinase (JNK) pathway has been shown to regulate mGluR-dependent nuclear transcription; however, the involvement of JNK in local, synaptic signaling has not been explored. Here, we show that JNK is both necessary and sufficient for mGluR-dependent expression of a subset of FMRP target proteins. In addition, JNK activity is basally elevated in fmr1 knockout mouse synapses, and blocking JNK activity reduces the over-expression of post-synaptic proteins in these mice. Together, these data suggest that JNK may be an important signaling mechanism downstream of mGluR stimulation, regulating FMRP-dependent protein synthesis. Furthermore, local, post-synaptic dysregulation of JNK activity may provide a viable target to ameliorate the deficits involved in FXS. Expression of many FMRP target proteins is enhanced in FXS. Here, we evaluated the role of JNKs in FXS. We found that JNK signaling is activated upon mGluR stimulation in wild-type neurons. Conversely, JNK activity is basally elevated in fmr1 knockout. Inhibiting JNK reduced the expression of FMRP target proteins and driving JNK activity increased the expression of these proteins.