EMAIL THIS PAGE TO A FRIEND
Innate immunity
MyD88 mediates the decision to die by apoptosis or necroptosis after UV irradiation.
PMID 24048771
Abstract
UV irradiation-induced cellular damage is classically associated with apoptosis and is known to result in systemic immunosuppression. How the decision to undergo apoptosis is made following UV is not fully understood. We hypothesize that a central mediator of TLR signaling, MyD88, determines cell fate after UV exposure. Survival after UV of immortalized bone marrow-derived macrophages (BMDM) and ex vivo peritoneal macrophages (PM) from MyD88 germline-deficient mice (MyD88(-/-)) was significantly higher than wild type (WT) PM. UV-induced apoptosis (DNA laddering) in PM and epidermis of MyD88(-/-) animals versus WT was decreased. In MyD88(-/-) PM, decreased cleavage of caspase 3, as well as pro-necroptotic protein, RIP1, and a significant increase in transcription and release of pro-inflammatory TNF-α, suggest that necroptosis, rather than apoptosis, has been initiated. Inxa0vivo studies confirm this hypothesis after UV, showing low apoptosis by TUNEL and inflammation in MyD88(-/-) skin sections. Considering that MyD88 participates in many TLR pathways, BMDM from TLR2(-/-), TLR4(-/-) and WT mice were compared for evidence of UV-induced apoptosis. Only TLR4(-/-) BMDM and PM had a similar phenotype to MyD88(-/-), suggesting that the TLR4-MyD88 axis importantly contributes to cell fate decision. Our study describes a new cellular consequence of MyD88 signaling after UV, and may provide rationale for therapies to mitigate UV-induced immunosuppression.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
GW23014 Anti-Actin, Cytoplasmic 1 antibody produced in chicken, affinity isolated antibody, buffered aqueous solution
HPA022034
Anti-MPRIP antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, Ab1
HPA022901
Anti-MPRIP antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, Ab2
SAB4500953
Anti-MPRIP antibody produced in rabbit, affinity isolated antibody
HPA046651
Anti-RALBP1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB1410141
Anti-RALBP1 antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
SAB2105823
Anti-RALBP1, (N-terminal) antibody produced in rabbit, affinity isolated antibody
R4277
Anti-RIP3 antibody produced in rabbit, ~1.0 mg/mL, affinity isolated antibody, buffered aqueous solution
PRS2283
Anti-RIP3 antibody produced in rabbit, affinity isolated antibody
GW21168 Anti-RIPK1 antibody produced in chicken, affinity isolated antibody, buffered aqueous solution
HPA015257
Anti-RIPK1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
AV02038
Anti-RIPK1 antibody produced in rabbit, affinity isolated antibody
SAB3500420
Anti-RIPK1 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
AV31513
Anti-RIPK3 antibody produced in rabbit, IgG fraction of antiserum
SAB2102009
Anti-RIPK3 antibody produced in rabbit, affinity isolated antibody, lyophilized powder
SAB2103903
Anti-RIPK3 antibody produced in rabbit, affinity isolated antibody
HPA055087
Anti-RIPK3 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB1401396
Anti-UQCRFS1 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
SAB1401397
Anti-UQCRFS1 antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
HPA041863
Anti-UQCRFS1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB2105078
Anti-UQCRFS1, (N-terminal) antibody produced in rabbit, affinity isolated antibody
11684809910
In Situ Cell Death Detection Kit, AP
WH0010928M2
Monoclonal Anti-RALBP1 antibody produced in mouse, clone 2A1, purified immunoglobulin, buffered aqueous solution
SAB1404880 Monoclonal Anti-RALBP1 antibody produced in mouse, clone 1C2-A10, purified immunoglobulin, buffered aqueous solution
SAB4100176
Monoclonal Anti-RALBP1 antibody produced in mouse, clone RALBP1-A192, culture supernatant