EMAIL THIS PAGE TO A FRIEND
Biochimica et biophysica acta
The role of abnormalities in the distal pathway of cholesterol synthesis in the Congenital Hemidysplasia with Ichthyosiform erythroderma and Limb Defects (CHILD) syndrome.
PMID 24060582
Abstract
CHILD syndrome (Congenital Hemidysplasia with Ichthyosiform erythroderma and Limb Defects) is a rare X-linked dominant ichthyotic disorder. CHILD syndrome results from loss of function mutations in the NSDHL gene, which leads to inhibition of cholesterol synthesis and accumulation of toxic metabolic intermediates in affected tissues. The CHILD syndrome skin is characterized by plaques topped by waxy scales and a variety of developmental defects in extracutaneous tissues, particularly limb hypoplasia or aplasia. Strikingly, these alterations are commonly segregated to either the right or left side of the body midline with little to no manifestations on the ipsilateral side. By understanding the underlying disease mechanism of CHILD syndrome, a pathogenesis-based therapy has been developed that successfully reverses the CHILD syndrome skin phenotype and has potential applications to the treatment of other ichthyoses. This article is part of a Special Issue entitled The Important Role of Lipids in the Epidermis and their Role in the Formation and Maintenance of the Cutaneous Barrier. Guest Editors: Kenneth R. Feingold and Peter Elias.