EMAIL THIS PAGE TO A FRIEND
The Science of the total environment
Analysis of U.S. soil lead (Pb) studies from 1970 to 2012.
PMID 24076506
Abstract
Although lead (Pb) emissions to the air have substantially decreased in the United States since the phase-out of leaded gasoline by 1995, amounts of lead in some soils remain elevated. Lead concentrations in residential and recreational soils are of concern because health effects have been associated with Pb exposure. Elevated soil Pb is especially harmful to young children due to their higher likelihood of soil ingestion. The purpose of this study is to create a comprehensive compilation of U.S. soil Pb data published from 1970 through 2012 as well as to analyze the collected data to reveal spatial and/or temporal soil Pb trends in the U.S. over the past 40 years. A total of 84 soil Pb studies across 62 U.S. cities were evaluated. Median soil Pb values from the studies were analyzed with respect to year of sampling, residential location type (e.g., urban, suburban), and population density. In aggregate, there was no statistically significant correlation between year and median soil Pb; however, within single cities, soil Pb generally declined over time. Our analysis shows that soil Pb quantities in city centers were generally highest and declined towards the suburbs and exurbs of the city. In addition, there was a statistically significant, positive relationship between median soil Pb and population density. In general, the trends examined here align with previously reported conclusions that soil Pb levels are higher in larger urban areas and Pb tends to remain in soil for long periods of time.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
GF32781999
Lead, microfoil, disks, 10mm, thinness 0.001μm, specific density 1.135μg/cm2, permanent mylar 3.5μm support, 99.99+%
Pb
GF64961163
Lead, microfoil, disks, 10mm, thinness 0.025μm, specific density 30.9μg/cm2, permanent mylar 3.5μm support, 99.99+%
Pb
GF13065218
Lead, microfoil, disks, 10mm, thinness 0.1μm, specific density 103.4μg/cm2, permanent mylar 3.5μm support, 99.99+%
Pb
GF47460753
Lead, microfoil, disks, 10mm, thinness 0.25μm, specific density 266.7μg/cm2, permanent mylar 3.5μm support, 99.99+%
Pb
GF89832033
Lead, microfoil, disks, 10mm, thinness 1.0μm, specific density 1047.8μg/cm2, permanent mylar 3.5μm support, 99.99+%
Pb
GF25514670
Lead, microfoil, disks, 25mm, thinness 1.0μm, specific density 1047.8μg/cm2, permanent mylar 3.5μm support, 99.99+%
Pb
GF10093362
Lead, rod, 1000mm, diameter 12.7mm, 99.95%
Pb
GF79893444
Lead, rod, 1000mm, diameter 19.0mm, 99.95%
Pb
GF92060489
Lead, rod, 1000mm, diameter 3.2mm, 99.95%
Pb
GF66906197
Lead, rod, 1000mm, diameter 6.35mm, 99.95%
Pb
GF05266672
Lead, rod, 100mm, diameter 19.0mm, 99.95%
Pb
GF55722359
Lead, rod, 100mm, diameter 50mm, 99.95%
Pb
GF07560435
Lead, rod, 100mm, diameter 6.35mm, 99.95%
Pb
GF79027847
Lead, rod, 200mm, diameter 12.7mm, 99.95%
Pb
GF58334711
Lead, rod, 200mm, diameter 19.0mm, 99.95%
Pb
GF45053883
Lead, rod, 200mm, diameter 6.35mm, 99.95%
Pb
GF91839145
Lead, rod, 500mm, diameter 19.0mm, 99.95%
Pb
GF67936483
Lead, rod, 500mm, diameter 3.2mm, 99.95%
Pb
GF31735245
Lead, rod, 500mm, diameter 6.35mm, 99.95%
Pb
GF65793482
Lead, rod, 50mm, diameter 50mm, 99.95%
Pb
GF52882533
Lead, tube, 100mm, outside diameter 6.78mm, inside diameter 4.7mm, wall thickness 1.04mm, 99.9%
Pb
GF10316742
Lead, tube, 200mm, outside diameter 6.78mm, inside diameter 4.7mm, wall thickness 1.04mm, 99.9%
Pb
GF74266061
Lead, microfoil, disks, 25mm, thinness 0.25μm, specific density 266.7μg/cm2, permanent mylar 3.5μm support, 99.99+%
Pb
GF86232225
Lead, rod, 100mm, diameter 12.7mm, 99.95%
Pb
GF99153085
Lead, rod, 200mm, diameter 50mm, 99.95%
Pb
GF99066190
Lead, tube, 1000mm, outside diameter 6.78mm, inside diameter 4.7mm, wall thickness 1.04mm, 99.9%
Pb
GF06993412
Lead, wire reel, 0.1m, diameter 1.0mm, as drawn, 99.99+%
Pb
GF97912985
Lead, wire reel, 0.5m, diameter 2.0mm, as drawn, 99.99+%
Pb
GF66008167
Lead, wire reel, 10m, diameter 0.25mm, as drawn, 99.95%
Pb
GF31296551
Lead, wire reel, 1m, diameter 0.25mm, as drawn, 99.95%
Pb