EMAIL THIS PAGE TO A FRIEND
American journal of physiology. Lung cellular and molecular physiology
Decreased Kv1.5 expression in intrauterine growth retardation rats with exaggerated pulmonary hypertension.
PMID 24077947
Abstract
Chronic hypoxia pulmonary hypertension (CH-PHT) in adulthood is likely to be of fetal origin following intrauterine growth retardation (IUGR). Oxygen (O₂)-sensitive voltage-gated potassium channels (Kv channels) in resistance pulmonary artery smooth muscle cells (PASMCs) play an important role in scaling pulmonary artery (PA) pressure. Expression and functional changes of Kv channels are determined, in part, by embryonic development. We hypothesized that O₂-sensitive Kv channels play an important role in exaggerated CH-PHT following IUGR. We established a rat model of IUGR by restricting maternal food during the entire pregnancy and exposed IUGR rats and their age-matched controls aged 12 wk to hypoxia for 2 wk. We found that hypoxia exposure significantly induced increased PA pressure and thicker smooth muscle layer in the IUGR group relative to controls. We compared the constriction of the resistance PA to inhibitors of K⁺ channels, 4-aminopyridine (4-AP), tetraethylammonium, and BaCl₂. Despite the thickness of the smooth muscle layer, the constriction to 4-AP was significantly reduced in the IUGR group exposed to hypoxia. Consistent with these changes in pulmonary vascular reactivity, 2 wk of hypoxia induced weaker 4-AP-sensitive Kv currents in a single IUGR PASMC. Moreover, after 2 wk of hypoxia, Kv1.5 expression in resistance PAs decreased significantly in the IUGR group. Overexpression of Kv1.5 in cultured PASMCs could offset hypoxia-induced cell proliferation and hypoxia-inhibited Kv currents in the IUGR group. These results suggest that the inhibited expression of Kv1.5 in PASMCs contribute to the development of exaggerated CH-PHT in IUGR rats during adulthood.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
HPA014864
Anti-KCNS3 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
449644
Barium chloride, anhydrous, beads, −10 mesh, 99.999% trace metals basis
BaCl2
449652
Barium chloride, anhydrous, beads, −10 mesh, 99.95% trace metals basis
BaCl2
202738
Barium chloride, 99.999% trace metals basis
BaCl2
B0750
Barium chloride dihydrate, ≥99%
BaCl2 · 2H2O
529591
Barium chloride dihydrate, ≥99.999% trace metals basis
BaCl2 · 2H2O
217565
Barium chloride dihydrate, ACS reagent, ≥99%
BaCl2 · 2H2O
11760
Barium chloride dihydrate, puriss. p.a., ACS reagent, ≥99.0% (KT)
BaCl2 · 2H2O
31125
Barium chloride dihydrate, puriss. p.a. plus, ACS reagent, reag. ISO, reag. Ph. Eur., ≥99%
BaCl2 · 2H2O
11722
Barium standard for AAS, ready-to-use, traceable to BAM, in H2O
BaCl2
G6539
Monoclonal Anti-Green Fluorescent Protein (GFP) antibody produced in mouse, clone GFP-20, ascites fluid
241059
Tetraethylammonium bromide, ReagentPlus®, 99%
C8H20BrN
140023
Tetraethylammonium bromide, reagent grade, 98%
C8H20BrN
86608
Tetraethylammonium bromide, for ion pair chromatography, ≥99.0%
C8H20BrN
113042
Tetraethylammonium chloride hydrate
C8H20ClN · xH2O
86636
Tetraethylammonium hydroxide solution, ~1.0 M (CH3CH2)4NOH in H2O, electrochemical grade
C8H21NO
86631
Tetraethylammonium hydroxide solution, ~25% in methanol (~1.5 M)
C8H21NO
86633
Tetraethylammonium hydroxide solution, ~40% in H2O
C8H21NO
86630
Tetraethylammonium hydroxide solution, technical, ~20% in H2O
C8H21NO
302929
Tetraethylammonium hydroxide solution, 35 wt. % in H2O
C8H21NO
177806
Tetraethylammonium hydroxide solution, 20 wt. % in H2O
C8H21NO
86640
Tetraethylammonium iodide, puriss., ≥98.5% (CHN)
C8H20IN
235938
Tetraethylammonium iodide, 98%
C8H20IN