EMAIL THIS PAGE TO A FRIEND
Iranian Red Crescent medical journal
A survey on the prevalence of Chlamydia trachomatis and Mycoplasma genitalium infections in symptomatic and asymptomatic men referring to urology clinic of labbafinejad hospital, tehran, iran.
PMID 24083010
Abstract
Chlamydia trachomatis and Mycoplasma genitalium infections are the most prevalent sexually transmitted bacterial infections in the world that cause urogenital infections in both men and women. It appears that infertility is a complication of these infections. This study was designed to estimate the prevalence of Chlamydia trachomatis and Mycoplasma genitalium in symptomatic and asymptomatic men and to assess risk factors associated with infection. Urine specimens were collected from 200 men; 100 of them were symptomatic and 100 asymptomatic. Samples were examined by PCR to detect the infections. C. trachomatis was detected in 20% of symptomatic and in 4% of asymptomatic men (P < 0.001). The prevalence of M. genitalium was revealed to be 12% and 2% in symptomatic and asymptomatic men, respectively (P < 0.01). Four of 100 men in the symptomatic group were infected with both organisms. C. trachomatis infection was associated with dysuria, urethral discharge, testicular swelling, and genital ulcer (P < 0.05). M. genitalium infection was related with dysuria, testis inflammation, pelvic pain and low educational level (P < 0.05). Furthermore, the prevalence of infections at ages 30-39 years was more than other ages. Considering the role of these bacteria in urogenital infections, a screening test is recommended. Since the PCR assay is a highly sensitive and specific assay for the detection of these bacteria in male urine specimens, it provides a noninvasive technique for routine screening.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
11699105001
PCR Buffer Without MgCl2, 10x concentrated