EMAIL THIS PAGE TO A FRIEND
Oxidative medicine and cellular longevity
Protective effects of extracts from Fructus rhodomyrti against oxidative DNA damage in vitro and in vivo.
PMID 24089629
Abstract
To evaluate the potential protective effects of extracts from Fructus rhodomyrti (FR) against oxidative DNA damage using a cellular system and the antioxidant ability on potassium bromate- (KBrO3-) mediated oxidative stress in rats. The effects of FR on DNA damage induced by hydrogen peroxide (H2O2) were evaluated by comet assay in primary spleen lymphocytes cultures. The effects of FR on the activities of SOD, CAT, and GPx and the levels of GSH, hydroperoxides, and 8-OHdG were determined in the plasma and tissues of rats treated with KBrO3. FR was shown to effectively protect against DNA damage induced by H2O2  in vitro, and the maximum protective effect was observed when FR was diluted 20 times. Endogenous antioxidant status, namely, the activities of SOD, CAT, and GPx and the levels of GSH were significantly decreased in the plasma, the liver, and the kidney of the KBrO3-treated rats, while the pretreatment of FR prevented the decreases of these parameters. In addition, the pretreatment of FR was also able to prevent KBrO3-induced increases in the levels of hydroperoxides and 8-OHdG in the plasma, the liver, and the kidney in rats. Our findings suggested that FR might act as a chemopreventive agent with antioxidant properties offering effective protection against oxidative DNA damage in a concentration-dependent manner in vitro and in vivo.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
730149
N-(3-Methoxypropyl)acrylamide, contains MEHQ as inhibitor, 95%
C7H13NO2