EMAIL THIS PAGE TO A FRIEND
British journal of cancer
Acquired resistance to temsirolimus in human renal cell carcinoma cells is mediated by the constitutive activation of signal transduction pathways through mTORC2.
PMID 24091619
Abstract
The objective of this study was to characterise the mechanism underlying acquired resistance to temsirolimus, an inhibitor of mammalian target of rapamycin (mTOR), in renal cell carcinoma (RCC). A parental human RCC cell line, ACHN (ACHN/P), was continuously exposed to increasing doses of up to 20 μM of temsirolimus, and a cell line resistant to temsirolimus (ACHN/R), showing a sixfold higher IC50 than that of ACHN/P, was developed. Following treatment with temsirolimus, phosphorylation of S6 kinase in ACHN/P was markedly inhibited, whereas there was no detectable expression of phosphorylated S6 in ACHN/R before and after temsirolimus treatment. However, AKT and p44/42 mitogen-activated protein kinase (MAPK) were constitutively phosphorylated even after temsirolimus treatment in ACHN/R, but not in ACHN/P. There was no significant difference between the sensitivities of ACHN/P and ACHN/R to KU0063794, a dual inhibitor of mTOR complex 1 (mTORC1) and mTORC2. Similar sensitivities to temsirolimus in ACHN/P and ACHN/R could be achieved by additional treatment with specific inhibitors of AKT- and MAPK-signaling pathways. The activation of signal transduction pathways via mTORC2, but not via mTORC1, may have an important role in the acquisition of a resistant phenotype to temsirolimus in RCC.