EMAIL THIS PAGE TO A FRIEND
Structure (London, England : 1993)
Crystal structure of human Seryl-tRNA synthetase and Ser-SA complex reveals a molecular lever specific to higher eukaryotes.
PMID 24095058
Abstract
Seryl-tRNA synthetase (SerRS), an essential enzyme for translation, also regulates vascular development. This "gain-of-function" has been linked to the UNE-S domain added to vertebrate SerRS during evolution. However, the significance of two insertions also specific to higher eukaryotic SerRS remains elusive. Here, we determined the crystal structure of human SerRS in complex with Ser-SA, an aminoacylation reaction intermediate analog, at 2.9xa0Å resolution. Despite a 70xa0Å distance, binding of Ser-SA in the catalytic domain dramatically leverages the position of Insertion I in the tRNA binding domain. Importantly, this leverage is specific to higher eukaryotes and not seen in bacterial, archaeal, and lower eukaryotic SerRSs. Deletion of Insertion I does not affect tRNA binding but instead reduce the catalytic efficiency of the synthetase. Thus, a long-range conformational and functional communication specific to higher eukaryotes is found in human SerRS, possibly to coordinate translation with vasculogenesis.