EMAIL THIS PAGE TO A FRIEND
Journal of gastroenterology
Epigenetic histone methylation regulates transforming growth factor β-1 expression following bile duct ligation in rats.
PMID 24097032
Abstract
Multiple mechanisms contribute to the liver fibrosis following cholestasis. Recent research has focused on the role of transforming growth factor β-1 (TGF-β1) in the progression of fibrosis. The aim of our study is to examine the role of epigenetic chromatin marks, such as histone H3 lysine methylation (H3Kme), in bile duct ligation (BDL)-induced TGF-β1 gene expression in rat liver. Time course of methylated-histone H3 and SET7/9 recruitment were determined by chromatin immunoprecipitation in livers from BDL rats on days 1, 4, 9 and 14. Levels of TGF-β1 and SET7/9 were determined by western blots. The effect of SET7/9 knockdown on BDL-induced expression of TGF-β1, serum enzymes and liver collagen content was studied in vivo. Results showed that BDL increased the expression of the TGF β-1. Increased levels of active chromatin marks (H3K4me1, H3K4me2, and H3K4me3) and decreased levels of repressive marks (H3K9me2 and H3K9me3) in TGF-β1 promoter accompanied the changes in expression of the TGF β-1. BDL also increased expression of the H3K4 methyltransferase SET7/9 and recruitment to the promoter. SET7/9 gene knockdown with siRNAs significantly attenuated BDL-induced TGF-β1 gene expression, serum enzymes and liver collagen content. Taken together, these results show the functional role of epigenetic chromatin histone H3Kme in BDL-induced TGF-β1 expression. Pharmacologic and other therapies that reverse these modifications could have potential hepatoprotective effects for BDL-induced cirrhosis.