EMAIL THIS PAGE TO A FRIEND
PLoS pathogens
Salmonella modulation of host cell gene expression promotes its intracellular growth.
PMID 24098123
Abstract
Salmonella Typhimurium has evolved a complex functional interface with its host cell largely determined by two type III secretion systems (T3SS), which through the delivery of bacterial effector proteins modulate a variety of cellular processes. We show here that Salmonella Typhimurium infection of epithelial cells results in a profound transcriptional reprogramming that changes over time. This response is triggered by Salmonella T3SS effector proteins, which stimulate unique signal transduction pathways leading to STAT3 activation. We found that the Salmonella-stimulated changes in host cell gene expression are required for the formation of its specialized vesicular compartment that is permissive for its intracellular replication. This study uncovers a cell-autonomous process required for Salmonella pathogenesis potentially opening up new avenues for the development of anti-infective strategies that target relevant host pathways.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SML0330
S3I-201, ≥97% (HPLC)
C16H15NO7S