PloS one
Mmp17b is essential for proper neural crest cell migration in vivo.
PMID 24098510
Abstract
The extracellular matrix plays a critical role in neural crest (NC) cell migration. In this study, we characterize the contribution of the novel GPI-linked matrix metalloproteinase (MMP) zebrafish mmp17b. Mmp17b is expressed post-gastrulation in the developing NC. Morpholino inactivation of mmp17b function, or chemical inhibition of MMP activity results in aberrant NC cell migration with minimal change in NC proliferation or apoptosis. Intriguingly, a GPI anchored protein with metalloproteinase inhibitor properties, Reversion-inducing-Cysteine-rich protein with Kazal motifs (RECK), which has previously been implicated in NC development, is expressed in close apposition to NC cells expressing mmp17b, raising the possibility that these two gene products interact. Consistent with this possibility, embryos silenced for mmp17b show defective development of the dorsal root ganglia (DRG), a crest-derived structure affected in RECK mutant fish sensory deprived (sdp). Taken together, this study has identified the first pair of MMP, and their putative MMP inhibitor RECK that functions together in NC cell migration.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A9563
4-Aminophenylmercuric acetate, ≥90% (titration)
C8H9HgNO2
M2699
Marimastat, ≥98% (HPLC)
C15H29N3O5