EMAIL THIS PAGE TO A FRIEND
Clinical genetics
Ethnic differences in GRHPR mutations in patients with primary hyperoxaluria type 2.
PMID 24116921
Abstract
The objective of this study was to investigate ethnic differences in the glyoxylate reductase/hydroxypyruvate reductase (GRHPR) gene in patients with primary hyperoxaluria type 2 (PH2). GRHPR was genotyped in Japanese patients with PH2 and all GRHPR mutations described to date were reviewed in terms of geographic and ethnic association. We identified a novel mutation, a two-nucleotide deletion (c.248_249delTG) in exon 3 creating a premature 'stop' at codon 91. Also, we found that the c.864_865delTG mutation was associated with the rs35891798 single-nucleotide polymorphism. The allelic frequencies of the c.103delG, c.494G>A, c.403_404+2 delAAGT, and c.864_865delTG mutations in PH2 patients were 37.8%, 15.6%, 10.0%, and 10.0%, respectively. All patients with the c.103delG mutation were Caucasian. Patients with the c.494G>A mutation and 78% (7/9) of those with the c.403_404+2 delAAGT mutation were from the Indian subcontinent, whereas those with the c.864_865delTG mutation were Chinese or Japanese. Molecular analysis of GRHPR of four Japanese PH2 patients identified a novel mutation (c.248_249delTG in exon 3). Caucasians with PH2 should be screened for the c.103delG mutation; patients from the Indian subcontinent for c.494G>A; and patients of East Asian origin (particularly) for c.864_865delTG. The prevalence of the latter mutation in PH2 patients from East Asia was 75.0%.