EMAIL THIS PAGE TO A FRIEND
Angewandte Chemie (International ed. in English)
Selective reduction of barbituric acids using SmI2/H2O: synthesis, reactivity, and structural analysis of tetrahedral adducts.
PMID 24123558
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
185698
Barbituric acid, ReagentPlus®, 99%
C4H4N2O3