EMAIL THIS PAGE TO A FRIEND
Critical care medicine
Inhaled nitric oxide does not reduce mortality in patients with acute respiratory distress syndrome regardless of severity: systematic review and meta-analysis.
PMID 24132038
Abstract
Treatment with inhaled nitric oxide improves oxygenation but not survival in mechanically ventilated patients with acute respiratory distress syndrome, but the effect may depend on the severity of hypoxemia. Our objective was to determine whether nitric oxide reduces hospital mortality in patients with severe acute respiratory distress syndrome (PaO2/FIO2 ≤ 100 mm Hg) but not in patients with mild-moderate acute respiratory distress syndrome (100 < PaO2/FIO2 ≤ 300 mm Hg) at the time of randomization. Data were collected from Medline, Embase, and Cochrane CENTRAL electronic databases (inception to May 2013); proceedings from five conferences (to May 2013); and trial registries (http://www.clinicaltrials.gov and http://www.controlled-trials.com). No language restrictions were applied. Two authors independently selected parallel-group randomized controlled trials comparing nitric oxide with control (placebo or no gas) in mechanically ventilated adults or postneonatal children with acute respiratory distress syndrome. Two authors independently extracted data from included trials. Trial investigators provided subgroup data. Meta-analyses used within-trial subgroups and random-effects models. Nine trials (n = 1,142 patients) met inclusion criteria. Overall methodological quality was good. Nitric oxide did not reduce mortality in patients with severe acute respiratory distress syndrome (risk ratio, 1.01 [95% CI, 0.78-1.32]; p = 0.93; n = 329, six trials) or mild-moderate acute respiratory distress syndrome (risk ratio, 1.12 [95% CI, 0.89-1.42]; p = 0.33; n = 740, seven trials). Risk ratios were similar between subgroups (interaction p = 0.53). There was no between-trial heterogeneity in any analysis (I = 0%). Varying the PaO2/FIO2 threshold between 70 and 200 mm Hg, in increments of 10 mm Hg, did not identify any threshold at which the nitric oxide-treated patients had lower mortality relative to controls. Nitric oxide does not reduce mortality in adults or children with acute respiratory distress syndrome, regardless of the degree of hypoxemia. Given the lack of related ongoing or recently completed randomized trials, new data addressing the effectiveness of nitric oxide in patients with acute respiratory distress syndrome and severe hypoxemia will not be available for the foreseeable future.