EMAIL THIS PAGE TO A FRIEND
Organic letters
A straightforward and highly diastereoselective access to functionalized monofluorinated cyclopropanes via a Michael initiated ring closure reaction.
PMID 24138105
Abstract
The synthesis of highly functionalized monofluorinated cyclopropanes based on a Michael Initiated Ring Closure (MIRC) reaction has been developed. The addition of quaternary ammonium salts derived from ethyl bromofluoroacetate on a panel of electron deficient alkenes followed by cyclization gave rise to an efficient access to monofluorinated cyclopropanes with good yields and remarkable diastereoselectivity.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
452114
Ethyl bromofluoroacetate, 97%
C4H6BrFO2