EMAIL THIS PAGE TO A FRIEND
Oncogene
PTEN functions as a melanoma tumor suppressor by promoting host immune response.
PMID 24141770
Abstract
Cancer cells acquire several traits that allow for their survival and progression, including the ability to evade the host immune response. However, the mechanisms by which cancer cells evade host immune responses remain largely elusive. Here we study the phenomena of immune evasion in malignant melanoma cells. We find that the tumor suppressor phosphatase and tensin homolog (PTEN) is an important regulator of the host immune response against melanoma cells. Mechanistically, PTEN represses the expression of immunosuppressive cytokines by blocking the phosphatidylinositide 3-kinase (PI3K) pathway. In melanoma cells lacking PTEN, signal transducer and activator of transcription 3 activates the transcription of immunosuppressive cytokines in a PI3K-dependent manner. Furthermore, conditioned media from PTEN-deficient, patient-derived short-term melanoma cultures and established melanoma cell lines blocked the production of the interleukin-12 (IL-12) in human monocyte-derived dendritic cells. Inhibition of IL-12 production was rescued by restoring PTEN or using neutralizing antibodies against the immunosuppressive cytokines. Furthermore, we report that PTEN, as an alternative mechanism to promote the host immune response against cancer cells, represses the expression of programmed cell death 1 ligand, a known repressor of the host immune response. Finally, to establish the clinical significance of our results, we analyzed malignant melanoma patient samples with or without brisk host responses. These analyses confirmed that PTEN loss is associated with a higher percentage of malignant melanoma samples with non-brisk host responses compared with samples with brisk host responses. Collectively, these results establish that PTEN functions as a melanoma tumor suppressor in part by regulating the host immune response against melanoma cells and highlight the importance of assessing PTEN status before recruiting melanoma patients for immunotherapies.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SAB4300337
Anti-PTEN (Ab-370) antibody produced in rabbit, affinity isolated antibody
SAB4300336
Anti-PTEN (Ab-380/382/383) antibody produced in rabbit, affinity isolated antibody
SAB2501614 Anti-PTEN antibody produced in goat, affinity isolated antibody, buffered aqueous solution
SAB1406331
Anti-PTEN antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
P7482
Anti-PTEN antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
SAB3500337
Anti-PTEN antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
EHU106441
MISSION® esiRNA, esiRNA human PTEN (esiRNA1)
SIHP0691 MISSION® Human Phosphatase, PTEN siRNA1, Nano Scale 250 pmol
SIHP0692 MISSION® Human Phosphatase, PTEN siRNA2, Nano Scale 250 pmol
SIHP0693 MISSION® Human Phosphatase, PTEN siRNA3, Nano Scale 250 pmol
WH0005728M1
Monoclonal Anti-PTEN antibody produced in mouse, clone 2G9, purified immunoglobulin, buffered aqueous solution
P3487
Monoclonal Anti-PTEN antibody produced in mouse, ~1 mg/mL, purified immunoglobulin, clone PTN-18, buffered aqueous solution
SAB1404272
Monoclonal Anti-PTEN antibody produced in mouse, clone 3E7, purified immunoglobulin, buffered aqueous solution
SAB4100580
Monoclonal Anti-PTEN antibody produced in mouse, clone PTEN-A552, culture supernatant
PLA0133
Rabbit anti-PTEN Antibody, Affinity Purified, Powered by Bethyl Laboratories, Inc.