EMAIL THIS PAGE TO A FRIEND
Clinics (Sao Paulo, Brazil)
Phytoestrogen α-zearalanol ameliorates memory impairment and neuronal DNA oxidation in ovariectomized mice.
PMID 24141843
Abstract
The aim of this study was to evaluate the effect of a novel phytoestrogen, α-Zearalanol, on Alzheimer's disease-related memory impairment and neuronal oxidation in ovariectomized mice. Female C57/BL6 mice were ovariectomized or received sham operations and treatment with equivalent doses of 17β-estradiol or α-Zearalanol for 8 weeks. Their spatial learning and memory were analyzed using the Morris water maze test. The antioxidant enzyme activities and reactive oxygen species generation, neuronal DNA oxidation, and MutT homolog 1 expression in the hippocampus were measured. Treatment with 17β-estradiol or α-Zearalanol significantly improved spatial learning and memory performance in ovariectomized mice. In addition, 17β-estradiol and α-Zearalanol attenuated the decrease in antioxidant enzyme activities and increased reactive oxygen species production in ovariectomized mice. The findings indicated a significant elevation in hippocampi neuronal DNA oxidation and reduction in MutT homolog 1 expression in estrogen-deficient mice, but supplementation with 17β-estradiol or α-Zearalanol efficaciously ameliorated this situation. These results demonstrate that α-Zearalanol is potentially beneficial for improving memory impairments and neuronal oxidation damage in a manner similar to that of 17β-estradiol. Therefore, the compound may be a potential therapeutic agent that can ameliorate neurodegenerative disorders related to estrogen deficiency.