EMAIL THIS PAGE TO A FRIEND
Journal of pediatric and adolescent gynecology
Isotretinoin: update on controversial issues.
PMID 24147278
Abstract
Acne vulgaris is a commonly seen dermatologic condition that plagues millions and can be very psychosocially disabling. Severe and recalcitrant acne is typically treated with isotretinoin. Isotretinoin is a synthetic vitamin A derivative that has been available since 1982.This therapeutic option has been the most effective at putting severe and recalcitrant acne vulgaris into remission. Despite its effectiveness,it has been associated with inflammatory bowel disease, depression, suicidality, and teratogenicity. We review the current literature on isotretinoin's role in inflammatory bowel disease, depression, and suicidality. In addition, we review whether or not the iPledge program has been successful at reducing pregnancy rates while on isotretinoin.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
1353500
Isotretinoin, United States Pharmacopeia (USP) Reference Standard
C20H28O2