EMAIL THIS PAGE TO A FRIEND
Leukemia
Functional impact of NOTCH1 mutations in chronic lymphocytic leukemia.
PMID 24170027
Abstract
The purpose of this study was to compare the expression and function of NOTCH1 in chronic lymphocytic leukemia (CLL) patients harboring a wild-type (WT) or mutated NOTCH1 gene. NOTCH1 mRNA and surface protein expression levels were independent of the NOTCH1 gene mutational status, consistent with the requirement for NOTCH1 signaling in this leukemia. However, compared with NOTCH1-WT CLL, mutated cases displayed biochemical and transcriptional evidence of an intense activation of the NOTCH1 pathway. In vivo, expression and activation of NOTCH1 was highest in CLL cells from the lymph nodes as confirmed by immunohistochemistry. In vitro, the NOTCH1 pathway was rapidly downregulated, suggesting that signaling relies upon micro-environmental interactions even in NOTCH1-mutated cases. Accordingly, co-culture of Jagged1(+) (the NOTCH1 ligand) nurse-like cells with autologous CLL cells sustained NOTCH1 activity over time and mediated CLL survival and resistance against pro-apoptotic stimuli, both abrogated when NOTCH1 signaling was pharmacologically switched off. Together, these results show that NOTCH1 mutations have stabilizing effects on the NOTCH1 pathway in CLL. Furthermore, micro-environmental interactions appear critical in activating the NOTCH1 pathway both in WT and mutated patients. Finally, NOTCH1 signals may create conditions that favor drug resistance, thus making NOTCH1 a potential molecular target in CLL.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SAB2700774
Anti-C21orf33 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
HPA018517
Anti-C21orf33 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB1401062
Anti-CD68 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
AV32372
Anti-HES1 antibody produced in rabbit, IgG fraction of antiserum
SAB2106243
Anti-HES1 antibody produced in rabbit, affinity isolated antibody
SAB1410659
Anti-HES1 antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
SAB4300493
Anti-MAPK3 (Ab-202) antibody produced in rabbit, affinity isolated antibody
SAB4300494
Anti-MAPK3 (Ab-204) antibody produced in rabbit, affinity isolated antibody
GW22781 Anti-MAPK3 antibody produced in chicken, affinity isolated antibody, buffered aqueous solution
SAB1412386
ANTI-MAPK3 antibody produced in mouse, clone 1E2, purified immunoglobulin, buffered aqueous solution
SAB2701851
Anti-mapk3 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB2701770
Anti-mapk3 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB2104574
Anti-MAPK3 antibody produced in rabbit, affinity isolated antibody
SAB2104732
Anti-MAPK3 antibody produced in rabbit, affinity isolated antibody
SAB4503422
Anti-MNK1 antibody produced in rabbit, affinity isolated antibody
SAB4503423
Anti-MNK1 antibody produced in rabbit, affinity isolated antibody
SAB4502019
Anti-NOTCH1 (Cleaved-Val1754) antibody produced in rabbit, affinity isolated antibody
N4788 Anti-NOTCH1 antibody produced in rabbit, ~1 mg/mL, affinity isolated antibody, buffered aqueous solution
SAB2101618
Anti-NOTCH1 antibody produced in rabbit, affinity isolated antibody
SAB4502389
Anti-p44 MAP Kinase antibody produced in rabbit, affinity isolated antibody
SAB4502390
Anti-p44 MAP Kinase antibody produced in rabbit, affinity isolated antibody
WH0008209M1
Monoclonal Anti-C21orf33 antibody produced in mouse, clone 1F5, purified immunoglobulin, buffered aqueous solution
AMAB90873
Monoclonal Anti-CD68 antibody produced in mouse, Prestige Antibodies® Powered by Atlas Antibodies, clone CL1338, purified immunoglobulin, buffered aqueous glycerol solution
AMAB90874
Monoclonal Anti-CD68 antibody produced in mouse, Prestige Antibodies® Powered by Atlas Antibodies, clone CL1346, purified immunoglobulin, buffered aqueous glycerol solution
WH0003280M1
Monoclonal Anti-HES1 antibody produced in mouse, clone 4D9, purified immunoglobulin, buffered aqueous solution
SAB3300102
Monoclonal Anti-HES1 antibody produced in mouse, clone 4H1, purified immunoglobulin, buffered aqueous glycerol solution
SAB1403945
Monoclonal Anti-HES1 antibody produced in mouse, clone 3A3, purified immunoglobulin, buffered aqueous solution
WH0005595M1
Monoclonal Anti-MAPK3 antibody produced in mouse, clone 3C9, purified immunoglobulin, buffered aqueous solution
SAB5300429
Monoclonal Anti-MAPK3 antibody produced in mouse, clone 1E5, ascites fluid
SAB4700742 Monoclonal Anti-Notch 1 antibody produced in mouse, clone mN1A, purified immunoglobulin, buffered aqueous solution