EMAIL THIS PAGE TO A FRIEND
Journal of medical virology
Apoptotic mediators in patients with severe and non-severe dengue from Brazil.
PMID 24170344
Abstract
Despite being the most significant arboviral disease worldwide, dengue has no antiviral treatment or reliable severity predictors. It has been shown that apoptotic cells from blood and tissues may be involved in the complex pathogenesis of dengue. However, very little is known about the interplay between proapoptotic and antiapoptotic mediators in this disease. Therefore, plasma levels of the three proapoptotic mediators Fas ligand (FasL), tumor necrosis factor-α (TNF-α), and TNF-related apoptosis-inducing ligand (TRAIL) were measured in dengue patients. Patients were classified according to the World Health Organization classification of dengue revised in 2009. Additionally, inhibitors of apoptosis protein (IAPs) were determined in plasma (Survivin) and peripheral blood mononuclear cells (PBMCs) lysates (cIAP-1, cIAP-2, XIAP). Levels of apoptotic proteins in plasma were correlated with counts of blood cells. FasL and TRAIL levels were elevated in dengue patients without warning signs when compared to patients with severe dengue and controls. Dengue patients with warning signs showed decreased levels of Survivin compared to patients with severe dengue and controls. TRAIL was inversely correlated with counts of lymphocyte subsets. In contrast, Survivin was positively correlated with leukocyte counts. There was a trend of elevated IAPs levels in PBMCs of patients with severe dengue. The results suggest a likely antiviral effect of TRAIL in dengue. It appears that TRAIL might be involved with apoptosis induction of lymphocytes, whereas IAPs might participate in protecting leukocytes from apoptosis. Further research is needed to explore the interactions between pro and antiapoptotic molecules and their implications in dengue pathogenesis.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
GW22783 Anti-CD14 antibody produced in chicken, affinity isolated antibody, buffered aqueous solution
SAB2500215
Anti-CD14 antibody produced in goat, affinity isolated antibody, buffered aqueous solution
SAB1405579
Anti-CD14 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
HPA001887
Anti-CD14 antibody produced in rabbit, Ab1, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
HPA002127
Anti-CD14 antibody produced in rabbit, Ab3, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
AMAB90897
Monoclonal Anti-CD14 antibody produced in mouse, Prestige Antibodies® Powered by Atlas Antibodies, clone CL1637, purified immunoglobulin, buffered aqueous glycerol solution
AMAB90898
Monoclonal Anti-CD14 antibody produced in mouse, Prestige Antibodies® Powered by Atlas Antibodies, clone CL1638, purified immunoglobulin, buffered aqueous glycerol solution
C7673
Monoclonal Anti-CD14 antibody produced in mouse, clone UCHM-1, purified immunoglobulin, buffered aqueous solution
SAB4700097
Monoclonal Anti-CD14 antibody produced in mouse, clone MEM-15, purified immunoglobulin, buffered aqueous solution
SAB4700103 Monoclonal Anti-CD14 antibody produced in mouse, clone MEM-18, purified immunoglobulin, buffered aqueous solution
SAB4700669 Monoclonal Anti-CD14 antibody produced in mouse, clone B-A8, purified immunoglobulin, buffered aqueous solution