EMAIL THIS PAGE TO A FRIEND
Journal of pharmaceutical and biomedical analysis
Determination of thioglycolic acid in cosmetics by capillary electrophoresis.
PMID 24184657
Abstract
A new and simple method for the accurate determination of thioglycolic acid (TGA) in cosmetics was developed using capillary electrophoresis (CE) with diode array detection at 236nm. The CE separation was performed on an uncoated fused silica capillary with a separation buffer solution containing 300mmolL(-1) tri-sodium phosphate and 0.5mmolL(-1) cetyltrimethylammonium bromide at a voltage of -5kV. Both the intra- and inter-day precisions of the method were 1.4%. The calibration curve between the corrected peak areas and the concentrations of the TGA was linear within the concentration range of 0.006-1.0mgmL(-1) with a correlation coefficient (r) of 0.9998. The limit of detection and limit of quantitation were 0.002mgmL(-1) (S/N=3) and 0.006mgmL(-1) (S/N=10), respectively. The average recoveries at the spiked levels of 0.125, 0.250 and 0.500mgmL(-1) were 96.9%, 102.3% and 94.0% with the relative standard derivations of 2.1%, 3.9% and 2.2%, respectively. The method was cross-validated by both high performance liquid chromatographic and ion chromatographic method. Eighty-five commercial depilatory creams and hair-treatment products were analyzed with satisfactory results.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
21250
Calcium thioglycolate trihydrate, crystallized, ≥98.0% (RT)
C2H3CaO2S · 3H2O