EMAIL THIS PAGE TO A FRIEND
Shanghai kou qiang yi xue = Shanghai journal of stomatology
[Influence of light irradiation mode on microhardness of dual cured resin cements].
PMID 24233200
Abstract
To investigate the influence of different light irradiation modes on microhardness of dual-curing resin cements. The cylindrical specimens (1 mm in thickness and 7.5 mm in diameter) of 5 self-adhesive resin cements (Biscem, Clearfil SA Cement, G-CEM LinkAce, Maxcem Elite and RelyX U100) and 2 universal resin cements(DUOLINK and Nexus 3) were irradiated respectively by a continuous mode (light-curing for 20 s) and a delayed mode (light-curing 5 s + auto-curing 60 s + light-curing 20 s), and then they were placed dry in a light-proof thermostat at 37 degrees centigrade. Surface microhardness of specimen was detected at 0.5, 24 h and 4 weeks post-irradiation. After that period of time, all specimens were soaked in ethanol for 24 h, their hardness were detected again, and the percentage of hardness reduction after soaking were calculated. Data were analyzed by SPSS 16.0 software package. The hardness of all resin cements under both irradiation modes increased mainly within 24 h post-irradiation. The irradiation mode did not affect the hardness of all resin cements 4 weeks after irradiation significantly (P>0.05), except for Biscem. However, after ethanol softening, 5 self-adhesive resin cements cured by the continuous mode had higher hardness reduction percentage than that cured by delayed mode (P<0.05); while hardness reduction percentage of 2 universal resin cements had no significant difference between continuous and delayed cure modes (P>0.05). The self-adhesive resin cements cured by delayed mode had better anti-softening ability. Supported by Nanjing Medical Science and Technology Development Fund (ZKX09035), Nanjing Science and Technology Development Fund (201001083) and Nanjing Medical Science and Technology Development Fund (YKK10125).
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
DUO92012
Duolink® In Situ Detection Reagents Brightfield
DUO92013
Duolink® In Situ Detection Reagents FarRed
DUO92014
Duolink® In Situ Detection Reagents Green
DUO92007
Duolink® In Situ Detection Reagents Orange
DUO92008
Duolink® In Situ Detection Reagents Red
DUO92106
Duolink® In Situ Orange Starter Kit Goat/Rabbit
DUO92104
Duolink® In Situ Orange Starter Kit Mouse/Goat
DUO92102
Duolink® In Situ Orange Starter Kit Mouse/Rabbit
DUO92006
Duolink® In Situ PLA® Probe Anti-Goat MINUS, Affinity purified Donkey anti-Goat IgG (H+L)
DUO92003
Duolink® In Situ PLA® Probe Anti-Goat PLUS, Affinity purified Donkey anti-Goat IgG (H+L)
DUO92004
Duolink® In Situ PLA® Probe Anti-Mouse MINUS, Affinity purified Donkey anti-Mouse IgG (H+L)
DUO92001
Duolink® In Situ PLA® Probe Anti-Mouse PLUS, Affinity purified Donkey anti-Mouse IgG (H+L)
DUO92005
Duolink® In Situ PLA® Probe Anti-Rabbit MINUS, Affinity purified Donkey anti-Rabbit IgG (H+L)
DUO92002
Duolink® In Situ PLA® Probe Anti-Rabbit PLUS, Affinity purified Donkey anti-Rabbit IgG (H+L)
DUO92010 Duolink® In Situ Probemaker MINUS
DUO92009 Duolink® In Situ Probemaker PLUS
DUO92105
Duolink® In Situ Red Starter Kit Goat/Rabbit
DUO92103
Duolink® In Situ Red Starter Kit Mouse/Goat
DUO92101
Duolink® In Situ Red Starter Kit Mouse/Rabbit