EMAIL THIS PAGE TO A FRIEND
Neuromuscular disorders : NMD
Novel TPM3 mutation in a family with cap myopathy and review of the literature.
PMID 24239060
Abstract
Cap myopathy is a rare congenital myopathy characterized by the presence of caps within muscle fibres and caused by mutations in ACTA1, TPM2 or TPM3. Thus far, only three cases with TPM3-related cap myopathy have been described. Here, we report on the first autosomal dominant family with cap myopathy in three-generations, caused by a novel heterozygous mutation in the alpha-tropomyosin-slow-encoding gene (TPM3; exon 4; c.445C>A; p.Leu149Ile). The three patients experienced first symptoms of muscle weakness in childhood and followed a slowly progressive course. They presented generalized hypotrophy and mild muscle weakness, elongated face, high arched palate, micrognathia, scoliosis and respiratory involvement. Intrafamilial variability of skeletal deformities, respiratory involvement and mild cardiac abnormalities was noted. Muscle MRI revealed a recognizable pattern of fatty muscle infiltration and masseter muscle hypertrophy. Subsarcolemmal caps were present in 6-10% of the fibres and immunoreactive with anti-tropomyosin antibodies. We conclude that the MRI-pattern of muscle involvement and the presence of masseter muscle hypertrophy in cap myopathy may guide molecular genetic diagnosis towards a mutation in TPM3. Regular respiratory examinations are important, even if patients have no anamnestic clues. We compare our findings to all cases of cap myopathy with identified mutations (n=11), thus far reported in the literature.