EMAIL THIS PAGE TO A FRIEND
Oncogene
Toward a therapeutic reduction of imatinib refractory myeloproliferative neoplasm-initiating cells.
PMID 24240679
Abstract
Myeloproliferative neoplasms (MPNs) such as chronic myelogenous (CML) and chronic myelomonocytic leukemias (CMML) are frequently induced by tyrosine kinase oncogenes. Although these MPNs are sensitive to tyrosine kinase inhibitors such as imatinib, patients often relapse upon withdrawal of therapy. We used a model of MPN, which is induced by co-expression of the oncoproteins HIP1/PDGFβR (H/P) and AML1/ETO from their endogenous loci, to examine the mechanisms of disease development and recurrence following imatinib withdrawal. Although the MPN displayed a full hematologic response to imatinib, 100% of the diseased mice relapsed upon drug withdrawal. MPN persistence was not due to imatinib resistance mutations in the H/P oncogene or massive gene expression changes. Within 1 week of imatinib treatment, more than 98% of gene expression changes induced by the oncogenes in isolated hematopoietic stem and progenitor cells (lineage(-)Sca-1(+)c-Kit(+) immunophenotype) normalized. Supplementation of imatinib with granulocyte colony-stimulating factor or arsenic trioxide reduced MPN-initiating cell frequencies and the combination of imatinib with arsenic trioxide cured a large fraction of mice with MPNs. In contrast, no mice in the imatinib-treated control cohorts were cured. These data suggest that treatment with a combination of arsenic trioxide and imatinib can eliminate refractory MPN-initiating cells and reduce disease relapse.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C3250000
Cyclophosphamide, European Pharmacopoeia (EP) Reference Standard
C7H15Cl2N2O2P · H2O