EMAIL THIS PAGE TO A FRIEND
Journal of medical microbiology
Molecular characterization of Escherichia coli isolates from patients with urinary tract infections in Palestine.
PMID 24243288
Abstract
Antibiotic resistance of Escherichia coli isolated from urinary tract infections (UTIs) is increasing worldwide. A total of 41 E. coli isolates were obtained from urine samples from hospitalized patients with a UTI in three hospitals in the northern districts of the West Bank, Palestine during March and June 2011. Resistance rates were: erythromycin (95 %), trimethoprim-sulfamethoxazole (59 %), ciprofloxacin (56 %), gentamicin (27 %), imipenem (22 %), amoxicillin (93 %), amoxicillin-clavulanic acid (32 %), ceftazidime (66 %) and cefotaxime (71 %). No meropenem-resistant isolates were identified in this study. Among the isolates, phylogenetic group B2 was observed in 13 isolates, D in 12 isolates, A in 11 isolates and B1 in five isolates. Thirty-five of the isolates were positive for an extended-spectrum β-lactamase phenotype. Among these isolates, the blaCTX-M gene was detected in 25, and eight harboured the blaTEM gene. None of the isolates contained the blaSHV gene. Transformation experiments indicated that some of the β-lactamase genes (i.e. blaCTX-M and blaTEM) with co-resistance to erythromycin and gentamicin were plasmid encoded and transmissible. Apart from this, enterobacterial repetitive intergenic consensus-PCR (ERIC-PCR) revealed that the 41 isolates were genetically diverse and comprised a heterogeneous population with 11 ERIC-PCR profiles at a 60 % similarity level.