EMAIL THIS PAGE TO A FRIEND
Biochimica et biophysica acta
Frequency decoding of calcium oscillations.
PMID 24269537
Abstract
Calcium (Ca(2+)) oscillations are ubiquitous signals present in all cells that provide efficient means to transmit intracellular biological information. Either spontaneously or upon receptor ligand binding, the otherwise stable cytosolic Ca(2+) concentration starts to oscillate. The resulting specific oscillatory pattern is interpreted by intracellular downstream effectors that subsequently activate different cellular processes. This signal transduction can occur through frequency modulation (FM) or amplitude modulation (AM), much similar to a radio signal. The decoding of the oscillatory signal is typically performed by enzymes with multiple Ca(2+) binding residues that diversely can regulate its total phosphorylation, thereby activating cellular program. To date, NFAT, NF-κB, CaMKII, MAPK and calpain have been reported to have frequency decoding properties. The basic principles and recent discoveries reporting frequency decoding of FM Ca(2+) oscillations are reviewed here. A limited number of cellular frequency decoding molecules of Ca(2+) oscillations have yet been reported. Interestingly, their responsiveness to Ca(2+) oscillatory frequencies shows little overlap, suggesting their specific roles in cells. Frequency modulation of Ca(2+) oscillations provides an efficient means to differentiate biological responses in the cell, both in health and in disease. Thus, it is crucial to identify and characterize all cellular frequency decoding molecules to understand how cells control important cell programs.