EMAIL THIS PAGE TO A FRIEND
Nanoscale
Programmed assembly of polymer-DNA conjugate nanoparticles with optical readout and sequence-specific activation of biorecognition.
PMID 24271079
Abstract
Soft micellar nanoparticles can be prepared from DNA conjugates designed to assemble via base pairing such that strands containing a polymer corona and a cholesterol tail generate controlled supramolecular architecture. Functionalization of one DNA conjugate strand with a biorecognition ligand results in shielding of the ligand when in the micelle, while encoding of the DNA sequences with overhangs allows supramolecular unpacking by addition of a complementary strand and sequence-specific unshielding of the ligand. The molecular assembly/disassembly and 'on-off' switch of the recognition signal is visualized by FRET pair signalling, PAGE and a facile turbidimetric binding assay, allowing direct and amplified readout of nucleic acid sequence recognition.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
31648
Sodium phosphotungstate tribasic hydrate, puriss. p.a.
Na3O40PW12 · xH2O