EMAIL THIS PAGE TO A FRIEND
Pharmaceutical development and technology
Medical-grade collagen peptide in injectables provides antioxidant protection.
PMID 24286241
Abstract
Medical-grade collagen peptide is used as an additive agent in pharmaceutical formulations; however, it is unknown as to whether the compound exerts antioxidant effects in vitro. In this study, we evaluated the antioxidant effects of medical-grade collagen peptide on reactive oxygen species such as hydroxyl radical, superoxide anion radical and singlet oxygen using electron spin resonance and spin trapping. We confirmed that medical-grade collagen peptide directly inhibited hydroxyl radical generated by the Fenton reaction or by ultraviolet irradiation of hydrogen peroxide, and singlet oxygen. In addition, an antioxidant effect of medical-grade collagen peptide on singlet oxygen was observed in peptide fractions 12-22. The total amount of antioxidant amino acids (Gly, Hyp, Glu, Ala, Cys, Met and His) constituted more than half of the total amino acids in these fractions. These results suggest that the observed antioxidant properties of medical-grade collagen peptide are due to the compound containing antioxidant amino acids. Medical-grade collagen peptide, which is used in pharmaceuticals, and especially in injectables, could provide useful antioxidant properties to protect the active ingredient from oxidation.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
115746
2,2,6,6-Tetramethyl-4-piperidinol, 98%
C9H19NO
F8263
Iron(II) sulfate heptahydrate, plant cell culture tested, ≥99%
FeO4S · 7H2O
F8633
Iron(II) sulfate heptahydrate, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99%
FeO4S · 7H2O
44970
Iron(II) sulfate heptahydrate, puriss. p.a., ACS reagent
FeO4S · 7H2O
215422
Iron(II) sulfate heptahydrate, ACS reagent, ≥99.0%
FeO4S · 7H2O
F7002
Iron(II) sulfate heptahydrate, ReagentPlus®, ≥99%
FeO4S · 7H2O
31236
Iron(II) sulfate heptahydrate, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., 99.0-103.4% (manganometric)
FeO4S · 7H2O
12354
Iron(II) sulfate heptahydrate, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, 99.5-104.5% (manganometric)
FeO4S · 7H2O
PHR1108
L-Histidine, Pharmaceutical Secondary Standard; Certified Reference Material
C6H9N3O2
73767
L-Histidine, certified reference material, TraceCERT®
C6H9N3O2
S3007
Sodium salicylate, ReagentPlus®, ≥99.5%
C7H5NaO3
S2679
Sodium salicylate, meets USP testing specifications
C7H5NaO3
71945
Sodium salicylate, puriss. p.a., ≥99.5% (NT)
C7H5NaO3
31493
Sodium salicylate, puriss. p.a., Reag. Ph. Eur., 99.5-101.0% (calc. to the dried substance)
C7H5NaO3
71943
Sodium salicylate, tested according to Ph.Eur.
C7H5NaO3
S0800000
Sodium salicylate, European Pharmacopoeia (EP) Reference Standard
C7H5NaO3