EMAIL THIS PAGE TO A FRIEND
Leukemia
Proteasome inhibitors exert cytotoxicity and increase chemosensitivity via transcriptional repression of Notch1 in T-cell acute lymphoblastic leukemia.
PMID 24301524
Abstract
The Notch signaling pathway has been recognized as a key factor for the pathogenesis of T-cell acute lymphoblastic leukemia (T-ALL), because of the high incidence of activating mutations of Notch1. Notch inhibition could serve as a new treatment strategy for T-ALL; however, the attempts to perturb Notch signaling pathways have been unsuccessful so far. In this study, we found that proteasome inhibitors exert cytotoxic effects on T-ALL cells with constitutive activation of Notch1 to a similar extent as myeloma cells. The proteasome inhibitor bortezomib repressed the transcription of Notch1 and downstream effectors including Hes1, GATA3, RUNX3 and nuclear factor-κB (NF-κB) (p65 and p50), coincided with downregulation of the major transactivator Sp1 and its dissociation from Notch1 promoter. Overexpression of the Notch1 intracellular domain (NICD) significantly ameliorated bortezomib-induced cytotoxicity against T-ALL cells. Drug combination studies revealed that bortezomib showed synergistic or additive effects with key drugs for the treatment of T-ALL such as dexamethasone (DEX), doxorubicin and cyclophosphamide, which were readily abolished by NICD overexpression. The synergy of bortezomib and DEX was confirmed in vivo using a murine xenograft model. Our findings provide a molecular basis and rationale for the inclusion of proteasome inhibitors in treatment strategies for T-ALL.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
AV37192
Anti-SP1 (AB1) antibody produced in rabbit, IgG fraction of antiserum
AV37193
Anti-SP1 (AB2) antibody produced in rabbit, IgG fraction of antiserum
SAB2500981
Anti-SP1 antibody produced in goat, affinity isolated antibody, buffered aqueous solution
SAB1412220
ANTI-SP1 antibody produced in mouse, clone 4H6, purified immunoglobulin, buffered aqueous solution
SAB1412261
ANTI-SP1 antibody produced in mouse, clone 4B11, purified immunoglobulin, buffered aqueous solution
SAB1412262
ANTI-SP1 antibody produced in mouse, clone 2G5, purified immunoglobulin, buffered aqueous solution
SAB1412263
ANTI-SP1 antibody produced in mouse, clone 3H7, purified immunoglobulin, buffered aqueous solution
SAB1412264
ANTI-SP1 antibody produced in mouse, clone 1G9, purified immunoglobulin, buffered aqueous solution
S9809 Anti-Sp1 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
HPA001853
Anti-SP1 antibody produced in rabbit, Ab1, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
AV43585
Anti-SP1 antibody produced in rabbit, affinity isolated antibody
HPA012292
Anti-SP1 antibody produced in rabbit, Ab2, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB4502836
Anti-SP1 antibody produced in rabbit, affinity isolated antibody
C3350000
Cytarabine, European Pharmacopoeia (EP) Reference Standard
C9H13N3O5
1162002
Cytarabine, United States Pharmacopeia (USP) Reference Standard
C9H13N3O5
PLA0307
Goat anti-Sp1 Antibody, Affinity Purified, Powered by Bethyl Laboratories, Inc.
WH0006667M2
Monoclonal Anti-SP1 antibody produced in mouse, clone 1A5, purified immunoglobulin, buffered aqueous solution
SAB1404397
Monoclonal Anti-SP1 antibody produced in mouse, clone 4C8, purified immunoglobulin, buffered aqueous solution
PLA0044
Rabbit anti-Sp1 Antibody, Affinity Purified, Powered by Bethyl Laboratories, Inc.